Order Entry
Canada
ContactUsLinkComponent
Anti-CLCF1 Rabbit Recombinant Polyclonal Antibody
Anti-CLCF1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CLCF1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77847-008
  • 77847-009
  • Antigen symbol:
  • CLCF1
  • CLCF1
  • Antigen synonyms:
  • B cell stimulating factor 3|B-cell-stimulating factor 3|BSF 3|BSF-3|BSF3|Cardiotrophin like cytokine|Cardiotrophin like cytokine factor 1|Cardiotrophin-like cytokine factor 1|CISS 2|CISS2|CLC|CLCF 1|CLCF1_HUMAN|Cold induced sweating syndrome 2|CRLF 1 associated cytokine like factor 1|CRLF1 associated cytokine like factor 1|Neurotrophin 1|Neurotrophin 1/B cell stimulating factor 3|Neurotrophin1|NNT 1|NNT-1|Novel neurotrophin 1|Novel neurotrophin-1|NR 6|NR6
  • B cell stimulating factor 3|B-cell-stimulating factor 3|BSF 3|BSF-3|BSF3|Cardiotrophin like cytokine|Cardiotrophin like cytokine factor 1|Cardiotrophin-like cytokine factor 1|CISS 2|CISS2|CLC|CLCF 1|CLCF1_HUMAN|Cold induced sweating syndrome 2|CRLF 1 associated cytokine like factor 1|CRLF1 associated cytokine like factor 1|Neurotrophin 1|Neurotrophin 1/B cell stimulating factor 3|Neurotrophin1|NNT 1|NNT-1|Novel neurotrophin 1|Novel neurotrophin-1|NR 6|NR6
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UBD9
  • Q9UBD9
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Cardiotrophin-like cytokine factor 1 protein (28-178AA)
  • Recombinant Human Cardiotrophin-like cytokine factor 1 protein (28-178AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together