Anti-CYP2F1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-923
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CYP2F1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P24903
- Antigen synonyms:CYP2F1Cytochrome P450 2F1|EC 1.14.14.1|CYPIIF1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cytochrome P450 2F1 protein (100-290AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: CYP2F1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human