Anti-PPP3CA Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: PPP3CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-359
- 77847-360
- Antigen symbol:
- PPP3CA
- PPP3CA
- Antigen synonyms:
- Alpha isoform formerly PPP2B|Calcineurin A alpha|Calcineurin A1|CalcineurinA|Calmodulin dependent calcineurin A subunit alpha isoform|Calmodulin-dependent calcineurin A subunit alpha isoform|CALN|CALNA 1|CALNA|CALNA1|CAM PRP catalytic subunit|CAM-PRP catalytic subunit|CCN 1|CCN1|CNA 1|CNA alpha|CNA|CNA1|PP2BA_HUMAN|PPP2B|Protein phosphatase 2B catalytic subunit|Protein phosphatase 3 (formerly 2B) catalytic subunit alpha isoform|Protein phosphatase 3 catalytic subunit alpha isoform Protein phosphatase 3 catalytic subunit alpha isozyme|Serine/threonine protein phosphatase 2B catalytic subunit alpha isoform|Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
- Alpha isoform formerly PPP2B|Calcineurin A alpha|Calcineurin A1|CalcineurinA|Calmodulin dependent calcineurin A subunit alpha isoform|Calmodulin-dependent calcineurin A subunit alpha isoform|CALN|CALNA 1|CALNA|CALNA1|CAM PRP catalytic subunit|CAM-PRP catalytic subunit|CCN 1|CCN1|CNA 1|CNA alpha|CNA|CNA1|PP2BA_HUMAN|PPP2B|Protein phosphatase 2B catalytic subunit|Protein phosphatase 3 (formerly 2B) catalytic subunit alpha isoform|Protein phosphatase 3 catalytic subunit alpha isoform Protein phosphatase 3 catalytic subunit alpha isozyme|Serine/threonine protein phosphatase 2B catalytic subunit alpha isoform|Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08209
- Q08209
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PPP3CA
- Fusion protein of Human PPP3CA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together