Anti-TRPV5 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TRPV5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77847-081
- 77847-080
- Antigen symbol:
- TRPV5
- TRPV5
- Antigen synonyms:
- Calcium transport protein 2|Calcium transporter 2|CAT 2|CAT2|ECAC 1|ECaC|ECaC1|Epithelial calcium channel 1|Osm 9 like TRP channel 3|Osm-9-like TRP channel 3|OTRPC 3|OTRPC3|Transient receptor potential cation channel subfamily V member 5|TRPV 5|TRPV5_HUMAN
- Calcium transport protein 2|Calcium transporter 2|CAT 2|CAT2|ECAC 1|ECaC|ECaC1|Epithelial calcium channel 1|Osm 9 like TRP channel 3|Osm-9-like TRP channel 3|OTRPC 3|OTRPC3|Transient receptor potential cation channel subfamily V member 5|TRPV 5|TRPV5_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQA5
- Q9NQA5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TRPV5
- Synthetic peptide of Human TRPV5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together