Anti-HMGN4 Rabbit Recombinant Polyclonal Antibody
77857-496
New Product
- Antigen symbol:HMGN4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O00479
- Antigen synonyms:HMG17L3|NHC|High mobility group nucleosome-binding domain-containing protein 4|Non-histone chromosomal protein HMG-17-like 3|Non-histone chromosomal protein
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human high mobility group nucleosome-binding domain-containing protein 4 protein (1-90AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HMGN4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human