Anti-MSMO1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: MSMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-704
- 77848-705
- Antigen symbol:
- MSMO1
- MSMO1
- Antigen synonyms:
- 1500001G16Rik|C 4 methyl sterol|C-4 methylsterol oxidase|C78600|DESP4|ERG25|Methylsterol monooxygenase 1|MGC104344|MSMO1|MSMO1_HUMAN|OTTHUMP00000219757|OTTHUMP00000219758|OTTHUMP00000219759|OTTHUMP00000219760|sterol C4 methyl oxidase like
- 1500001G16Rik|C 4 methyl sterol|C-4 methylsterol oxidase|C78600|DESP4|ERG25|Methylsterol monooxygenase 1|MGC104344|MSMO1|MSMO1_HUMAN|OTTHUMP00000219757|OTTHUMP00000219758|OTTHUMP00000219759|OTTHUMP00000219760|sterol C4 methyl oxidase like
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15800
- Q15800
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MSMO1
- Synthetic peptide of Human MSMO1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together