Anti-CD79B Rabbit Recombinant Polyclonal Antibody
77852-741
New Product
- Antigen symbol:CD79B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P40259
- Antigen synonyms:AGM6|B cell antigen receptor complex associated protein beta chain|B cell specific glycoprotein B29|B-cell antigen receptor complex-associated protein beta chain|B-cell-specific glycoprotein B29|B29|B29/Ig-beta/CD 79b|CD79b antigen (immunoglobulin associated beta)|CD79b antigen|CD79b molecule|CD79b molecule immunoglobulin associated beta|CD79b protein|CD79B_HUMAN|Ig beta|Ig-beta|IGB|Igbeta|Immunoglobulin associated B29|Immunoglobulin associated B29 protein|Immunoglobulin associated beta|Immunoglobulin associated protein|Immunoglobulin-associated B29 protein|MGC108607
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human B-cell antigen receptor complex-associated protein beta chain protein (29-159AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: CD79B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human