Anti-HSD11B1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-671
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HSD11B1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:P28845
- Antigen synonyms:11 beta HSD 1|11 beta HSD1|11 beta hydroxysteroid dehydrogenase 1|11 DH|11-beta hydroxysteroid dehydrogenase|type 1|11-beta-HSD1|11-beta-hydroxysteroid dehydrogenase 1|11-DH|11DH|Corticosteroid 11 beta dehydrogenase isozyme 1|Corticosteroid 11-beta-dehydrogenase isozyme 1|CORTRD2|DHI1_HUMAN|HDL|HSD 11|HSD11|HSD11B|HSD11L|Hydroxysteroid (11 beta) dehydrogenase|Hydroxysteroid (11 beta) dehydrogenase 1|MGC13539|SDR26C1|short chain dehydrogenase/reductase family 26C|member 1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Corticosteroid 11-beta-dehydrogenase isozyme 1 protein (25-292AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: HSD11B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human