Anti-MDH1 Rabbit Recombinant polyclonal Antibody
77859-602
New Product
- Antigen symbol:MDH1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P40925
- Antigen synonyms:cytoplasmic|Cytosolic malate dehydrogenase|Diiodophenylpyruvate reductase|Malate dehydrogenase 1, NAD (soluble)|Malate dehydrogenase|Malate dehydrogenase cytoplasmic|MDH s|MDHA|MDHC_HUMAN|MDHs|MGC:1375|MOR2|Soluble malate dehydrogenase
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Malate dehydrogenase, cytoplasmic protein (1-334AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:200-1:500, IF:1:50-1:200
Type:
Antigen: MDH1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse