Anti-AP3M2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77851-227
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:AP3M2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P53677
- Antigen synonyms:AP3M2|AP-3 complex subunit mu-2|Adaptor-related protein complex 3 subunit mu-2|Clathrin assembly protein assembly protein complex 3 mu-2 medium chain|Clathrin coat assembly protein AP47 homolog 2|Clathrin coat-associated protein AP47 homolog 2|Golgi adaptor AP-1 47 kDa protein homolog 2|HA1 47 kDa subunit homolog 2|Mu3B-adaptin|P47B
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-3 complex subunit mu-2 protein (1-273AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AP3M2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human