Anti-CALB1 Rabbit Recombinant Polyclonal Antibody
77852-274
New Product
- Antigen symbol:CALB1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P05937
- Antigen synonyms:avian-type|CAB27|CALB 1|CALB|CALB1|CALB1_HUMAN|Calbindin 1 28kDa|Calbindin|Calbindin D28|D 28K|D-28K|D28K|OTTHUMP00000166027|OTTHUMP00000225441|RTVL H protein|Vitamin D dependent calcium binding protein|Vitamin D dependent calcium binding protein avian type|Vitamin D-dependent calcium-binding protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Calbindin protein (2-261AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:1000 to 1:2000, IF: 1:200 to 1:500
Type:
Antigen: CALB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human