Anti-AURKC Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-699
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:AURKC
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UQB9
- Antigen synonyms:AIE 2|Aie1|AIE2|AIK 3|AIK3|ARK 3|ARK-3|ARK3|Aur C|AurC|Aurkc|AURKC_HUMAN|aurora 3|Aurora C|Aurora kinase C|aurora related kinase 3|Aurora-related kinase 3|Aurora/Ipl1 related kinase 3|Aurora/Ipl1-related kinase 3|Aurora/Ipl1/Eg2 protein 2|EC 2.7.11.1|Serine threonine protein kinase 13|serine/threonine kinase 13 (aurora/IPL1 like)|serine/threonine protein kinase aurora C|Serine/threonine-protein kinase 13|Serine/threonine-protein kinase aurora-C|SPGF5|STK13
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aurora kinase C protein (12-143AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: AURKC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human