Anti-MAST1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MAST1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-480
- 77846-481
- Antigen symbol:
- MAST1
- MAST1
- Antigen synonyms:
- 9430008B02Rik|KIAA0973|MAST 1|MAST1_HUMAN|Microtubule associated serine/threonine kinase 1|Microtubule associated serine/threonine protein kinase 1|Microtubule-associated serine/threonine-protein kinase 1|SAST 124|SAST 170|SAST|SAST170|Syntrophin associated serine/threonine kinase|Syntrophin associated serine/threonine protein kinase|Syntrophin-associated serine/threonine-protein kinase
- 9430008B02Rik|KIAA0973|MAST 1|MAST1_HUMAN|Microtubule associated serine/threonine kinase 1|Microtubule associated serine/threonine protein kinase 1|Microtubule-associated serine/threonine-protein kinase 1|SAST 124|SAST 170|SAST|SAST170|Syntrophin associated serine/threonine kinase|Syntrophin associated serine/threonine protein kinase|Syntrophin-associated serine/threonine-protein kinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2H9
- Q9Y2H9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Microtubule-associated serine/threonine-protein kinase 1 protein (687-879AA)
- Recombinant Human Microtubule-associated serine/threonine-protein kinase 1 protein (687-879AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together