Anti-EIF4EBP3 Rabbit Recombinant Polyclonal Antibody
77854-634
New Product
- Antigen symbol:EIF4EBP3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60516
- Antigen synonyms:EIF4EBP3Eukaryotic translation initiation factor 4E-binding protein 3|4E-BP3|eIF4E-binding protein 3
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Eukaryotic translation initiation factor 4E-binding protein 3 protein (1-100AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EIF4EBP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human