Anti-CD33 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-649
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CD33
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P20138
- Antigen synonyms:CD 33|CD33|CD33 antigen (gp67)|CD33 antigen|CD33 molecule|CD33_HUMAN|FLJ00391|gp67|My9|Myeloid cell surface antigen CD33|Myeloid cell surface antigen CD33 precursor|Myeloid differentiation antigen CD33|p67|Sialic acid binding Ig like lectin 3|Sialic acid binding immunoglobulin like lectin 3|Sialic acid-binding Ig-like lectin 3|SIGLEC 3|Siglec-3|SIGLEC3
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Myeloid cell surface antigen CD33 protein (49-259AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CD33
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human