Anti-GNMT Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: GNMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77847-880
- 77847-879
- Antigen symbol:
- GNMT
- GNMT
- Antigen synonyms:
- EC 2.1.1.20|Epididymis secretory sperm binding protein Li 182mP|Glycine N methyltransferase|Glycine N-methyltransferase|GNMT_HUMAN|HEL S 182mP|OTTHUMP00000016412
- EC 2.1.1.20|Epididymis secretory sperm binding protein Li 182mP|Glycine N methyltransferase|Glycine N-methyltransferase|GNMT_HUMAN|HEL S 182mP|OTTHUMP00000016412
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14749
- Q14749
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human GNMT
- Fusion protein of Human GNMT
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together