Anti-FKBP8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:1000 to 1:5000, IHC: 1:100 to 1:300
Type:
Antigen: FKBP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77849-816
- 77849-817
- Antigen symbol:
- FKBP8
- FKBP8
- Antigen synonyms:
- 38 kDa FK506-binding protein| 38 kDa FKBP| 38kDa| FK506 binding protein 8| FK506 binding protein 8, 38kDa| FK506 binding protein, 38kDa| FK506-binding protein 8| FKBP 38| FKBP-38| FKBP-8| FKBP8_HUMAN| FKBPR38| hFKBP38| Peptidyl-prolyl cis-trans isomerase PPIase Rotamase| Sam11
- 38 kDa FK506-binding protein| 38 kDa FKBP| 38kDa| FK506 binding protein 8| FK506 binding protein 8, 38kDa| FK506 binding protein, 38kDa| FK506-binding protein 8| FKBP 38| FKBP-38| FKBP-8| FKBP8_HUMAN| FKBPR38| hFKBP38| Peptidyl-prolyl cis-trans isomerase PPIase Rotamase| Sam11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14318
- Q14318
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human FKBP8
- Fusion protein of Human FKBP8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together