Anti-NQO1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:10000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77843-727
- 77843-726
- Antigen symbol:
- NQO1
- NQO1
- Antigen synonyms:
- Azoreductase|Cytochrome b 5 reductase|DHQU|DIA 4|DIA4|Diaphorase (NADH/NADPH) (cytochrome b 5 reductase)|Diaphorase (NADH/NADPH)|Diaphorase 4|Dioxin inducible 1|DT diaphorase|DT-diaphorase|DTD|Menadione reductase|NAD(P)H dehydrogenase [quinone] 1|NAD(P)H dehydrogenase quinone 1|NAD(P)H menadione oxidoreductase 1 dioxin inducible|NAD(P)H quinone dehydrogenase 1|NAD(P)H: menadione oxidoreductase 1 dioxin inducible 1|NAD(P)H:menadione oxidoreductase 1|NAD(P)H:Quinone acceptor oxidoreductase type 1|NAD(P)H:quinone oxidoreductase 1|NAD(P)H:quinone oxireductase|NMOR 1|NMOR I|NMOR1|NMORI|NQO 1|NQO1_HUMAN|Phylloquinone reductase|QR 1|QR1|Quinone reductase 1
- Azoreductase|Cytochrome b 5 reductase|DHQU|DIA 4|DIA4|Diaphorase (NADH/NADPH) (cytochrome b 5 reductase)|Diaphorase (NADH/NADPH)|Diaphorase 4|Dioxin inducible 1|DT diaphorase|DT-diaphorase|DTD|Menadione reductase|NAD(P)H dehydrogenase [quinone] 1|NAD(P)H dehydrogenase quinone 1|NAD(P)H menadione oxidoreductase 1 dioxin inducible|NAD(P)H quinone dehydrogenase 1|NAD(P)H: menadione oxidoreductase 1 dioxin inducible 1|NAD(P)H:menadione oxidoreductase 1|NAD(P)H:Quinone acceptor oxidoreductase type 1|NAD(P)H:quinone oxidoreductase 1|NAD(P)H:quinone oxireductase|NMOR 1|NMOR I|NMOR1|NMORI|NQO 1|NQO1_HUMAN|Phylloquinone reductase|QR 1|QR1|Quinone reductase 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15559
- P15559
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human NQO1
- Synthetic peptide of Human NQO1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together