Anti-ECEL1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-476
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ECEL1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O95672
- Antigen synonyms:ECEL1_HUMAN|Endothelin-converting enzyme-like 1|Xce protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Endothelin-converting enzyme-like 1 protein (425-775AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ECEL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human