Anti-HINT1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:100 to 1:300
Type:
Antigen: HINT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77847-652
- 77847-651
- Antigen symbol:
- HINT1
- HINT1
- Antigen synonyms:
- Adenosine 5' monophosphoramidase|Adenosine 5''-monophosphoramidase|HINT 1|HINT1_HUMAN|Histidine triad nucleotide binding protein 1|Histidine triad nucleotide-binding protein 1|PKCI 1|PKCI-1|PKCI1|PRKCNH1|Protein kinase C inhibitor 1|Protein kinase C interacting protein 1|Protein kinase C-interacting protein 1
- Adenosine 5' monophosphoramidase|Adenosine 5''-monophosphoramidase|HINT 1|HINT1_HUMAN|Histidine triad nucleotide binding protein 1|Histidine triad nucleotide-binding protein 1|PKCI 1|PKCI-1|PKCI1|PRKCNH1|Protein kinase C inhibitor 1|Protein kinase C interacting protein 1|Protein kinase C-interacting protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49773
- P49773
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human HINT1
- Synthetic peptide of Human HINT1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together