Anti-EGFL7 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EGFL7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-853
- 77843-854
- Antigen symbol:
- EGFL7
- EGFL7
- Antigen synonyms:
- EGF like domain 7|EGF like domain containing protein 7|EGF like domain multiple 7|EGF-like protein 7|EGFL 7|EGFL7_HUMAN|Epidermal growth factor like domain protein 7|Epidermal growth factor-like protein 7|MEGF 7|MEGF7|MGC111117|Multiple EGF like domain protein 7|Multiple EGF-like domains protein 7|Multiple epidermal growth factor like domain protein 7|Multiple epidermal growth factor-like domains protein 7|NEU1|NEU1 protein|NOTCH4 like protein|NOTCH4-like protein|RP11 251M1.2|UNQ187/PRO1449|Vascular endothelial statin|VE statin|VE-statin|ZNEU 1|ZNEU1
- EGF like domain 7|EGF like domain containing protein 7|EGF like domain multiple 7|EGF-like protein 7|EGFL 7|EGFL7_HUMAN|Epidermal growth factor like domain protein 7|Epidermal growth factor-like protein 7|MEGF 7|MEGF7|MGC111117|Multiple EGF like domain protein 7|Multiple EGF-like domains protein 7|Multiple epidermal growth factor like domain protein 7|Multiple epidermal growth factor-like domains protein 7|NEU1|NEU1 protein|NOTCH4 like protein|NOTCH4-like protein|RP11 251M1.2|UNQ187/PRO1449|Vascular endothelial statin|VE statin|VE-statin|ZNEU 1|ZNEU1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHF1
- Q9UHF1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Epidermal growth factor-like protein 7 protein (73-198AA)
- Recombinant human Epidermal growth factor-like protein 7 protein (73-198AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together