Anti-CDKN2B Rabbit Recombinant Polyclonal Antibody (Biotin)
77852-892
New Product
- Antigen symbol:CDKN2B
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P42772
- Antigen synonyms:CDK inhibitory protein|CDK4B Inhibitor|Cdkn2b|CDN2B_HUMAN|Cyclin dependent kinase 4 inhibitor B|Cyclin Dependent Kinase Inhibitor 2B|Cyclin dependent kinase inhibitor 2B p15 inhibits CDK4|Cyclin dependent kinases 4 and 6 binding protein|Cyclin-dependent kinase 4 inhibitor B|INK4B|MTS 2|MTS-2|MTS2|Multiple tumor suppressor 2|Multiple Tumor Supressor 2|OTTHUMP00000021154|OTTHUMP00000021155|p14 CDK inhibitor|p14 INK4b|p14-INK4b|P15|p15 CDK inhibitor|p15 inhibits CDK4|p15 INK4b|p15-INK4b|p15INK4B|TP 15|TP15
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cyclin-dependent kinase 4 inhibitor B protein (1-138AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: CDKN2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human