Anti-MBIP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: MBIP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-472
- 77843-471
- Antigen symbol:
- MBIP
- MBIP
- Antigen synonyms:
- MAP3K12 binding inhibitory protein 1|MAP3K12-binding inhibitory protein 1|MAPK upstream kinase binding inhibitory protein|MBIP1_HUMAN|MUK-binding inhibitory protein|OTTHUMP00000178844|MUK binding inhibitory protein|Mbip|MAPK upstream kinase-binding inhibitory protein
- MAP3K12 binding inhibitory protein 1|MAP3K12-binding inhibitory protein 1|MAPK upstream kinase binding inhibitory protein|MBIP1_HUMAN|MUK-binding inhibitory protein|OTTHUMP00000178844|MUK binding inhibitory protein|Mbip|MAPK upstream kinase-binding inhibitory protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NS73
- Q9NS73
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human MAP3K12-binding inhibitory protein 1 protein (91-241AA)
- Recombinant Human MAP3K12-binding inhibitory protein 1 protein (91-241AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together