Anti-DSP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77854-395
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:DSP
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P15924
- Antigen synonyms:Desmoplakin|DCWHKTA|Desmoplakin (DPI DPII)|DP II|Desmoplakin II|KPPS2|DP|PPKS2|DSP|DPII|PPKS 2|250/210 kDa paraneoplastic pemphigus antigen|Desmoplakin I|DPI|DESP_HUMAN|DP I
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Desmoplakin protein (78-300AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: DSP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human