Anti-CHST10 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77853-147
New Product
- Antigen symbol:CHST10
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O43529
- Antigen synonyms:CHST10Carbohydrate sulfotransferase 10|EC 2.8.2.-|HNK-1 sulfotransferase|HNK-1ST|HNK1ST|HuHNK-1ST
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Carbohydrate sulfotransferase 10 protein (28-356AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: CHST10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human