Anti-PIH1D1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77843-568
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PIH1D1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NWS0
- Antigen synonyms:NOP17|Nucleolar protein 17 homolog|PIH1 domain containing 1|PIH1 domain containing protein 1|PIH1 domain-containing protein 1|PIHD1_HUMAN
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human PIH1 domain-containing protein 1 protein (1-290AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PIH1D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human