Human Recombinant MICA (from HEK293 Cells)
Partial length Human MICA, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Shipped at 4 °C. Lyophilized: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q29983
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:MHC class I polypeptide-related sequence A|MIC-A|PERB11.1
- Protein/peptide name:MICA
- Purity:>95% by SDS-PAGE and Coomassie blue staining.
- Molecular weight:The protein has a predicted molecular mass of 33.5 kDa after removal of the signal peptide. The apparent molecular mass of MICA-His is approximately 55 - 70 kDa due to glycosylation.
- Sequence:MICA (Glu24-Trp307) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:ELISA, SDS-PAGE
Frequently Bought Together