Anti-RTN4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:50 to 1:200
Type:
Antigen: RTN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-883
- 77847-884
- Antigen symbol:
- RTN4
- RTN4
- Antigen synonyms:
- 1110020G17Rik|AA407876|AA409940|AA960376|ASY|C130026I10Rik|Foocen|Glut4 vesicle 20 kDa protein|Human NogoA|Kiaa0886|KIAA4153|MGC116054|MGC139261|mKIAA0886|mKIAA4153|My043 protein|Nbla00271|Nbla10545|Neurite growth inhibitor 220|Neurite Growth Inhibitor 220, included|Neurite outgrowth inhibitor|Neuroendocrine-specific protein|Neuroendocrine-specific protein C homolog|NI-250|NI220/250|Nogo A|NOGO|Nogo B|Nogo C|Nogo protein|NOGOC|NSP|NSP-CL|rat N|Reticulon 4|Reticulon 5|Reticulon-4|Reticulon-5|RTN X|RTN-x|Rtn4|Rtn4 reticulon 4|RTN4-A|RTN4-B1|RTN4-B2|RTN4-C|RTN4_HUMAN|Vp20
- 1110020G17Rik|AA407876|AA409940|AA960376|ASY|C130026I10Rik|Foocen|Glut4 vesicle 20 kDa protein|Human NogoA|Kiaa0886|KIAA4153|MGC116054|MGC139261|mKIAA0886|mKIAA4153|My043 protein|Nbla00271|Nbla10545|Neurite growth inhibitor 220|Neurite Growth Inhibitor 220, included|Neurite outgrowth inhibitor|Neuroendocrine-specific protein|Neuroendocrine-specific protein C homolog|NI-250|NI220/250|Nogo A|NOGO|Nogo B|Nogo C|Nogo protein|NOGOC|NSP|NSP-CL|rat N|Reticulon 4|Reticulon 5|Reticulon-4|Reticulon-5|RTN X|RTN-x|Rtn4|Rtn4 reticulon 4|RTN4-A|RTN4-B1|RTN4-B2|RTN4-C|RTN4_HUMAN|Vp20
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQC3
- Q9NQC3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human RTN4
- Synthetic peptide of Human RTN4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together