Anti-ELF5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ELF5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-263
- 77844-262
- Antigen symbol:
- ELF5
- ELF5
- Antigen synonyms:
- E74 like factor 5 (ets domain transcription factor)|E74 like factor 5|E74-like factor 5|ELF 5|ELF5_HUMAN|Epithelium restricted ESE 1 related Ets factor|Epithelium restricted ESE1 related Ets factor|Epithelium specific Ets transcription factor 2|Epithelium-restricted ESE-1-related Ets factor|Epithelium-specific Ets transcription factor 2|ESE 2|ESE-2|ESE2|Ets domain TF|Ets domain transcription factor|ETS related transcription factor Elf 5|ETS related transcription factor ETS-related transcription factor Elf-5
- E74 like factor 5 (ets domain transcription factor)|E74 like factor 5|E74-like factor 5|ELF 5|ELF5_HUMAN|Epithelium restricted ESE 1 related Ets factor|Epithelium restricted ESE1 related Ets factor|Epithelium specific Ets transcription factor 2|Epithelium-restricted ESE-1-related Ets factor|Epithelium-specific Ets transcription factor 2|ESE 2|ESE-2|ESE2|Ets domain TF|Ets domain transcription factor|ETS related transcription factor Elf 5|ETS related transcription factor ETS-related transcription factor Elf-5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKW6
- Q9UKW6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human ETS-related transcription factor Elf-5 protein (1-170AA)
- Recombinant Human ETS-related transcription factor Elf-5 protein (1-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together