Anti-PITX3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-731
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PITX3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:O75364
- Antigen synonyms:Homeobox protein PITX 3|Homeobox protein MGC12766|Paired like homeodomain transcription factor 3|Paired-like homeodomain transcription factor 3|Pituitary homeobox 3|PITX 3|PITX3_HUMAN|PTX 3|PTX3
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Pituitary homeobox 3 protein (10-200AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PITX3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse