Anti-FUT1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: FUT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-017
- 77848-016
- Antigen symbol:
- FUT1
- FUT1
- Antigen synonyms:
- 2)FT 1| 2-alpha-L-fucosyltransferase| Alpha (1 2) fucosyltransferase| Alpha(1 2)FT 1| Alpha(1| Blood group H alpha 2-fucosyltransferase| fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase)| Fucosyltransferase 1| FUT1| FUT1_HUMAN| Galactoside 2 alpha L fucosyltransferase| Galactoside 2-alpha-L-fucosyltransferase 1| GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1| H| HH| HSC| Para Bombay phenotype
- 2)FT 1| 2-alpha-L-fucosyltransferase| Alpha (1 2) fucosyltransferase| Alpha(1 2)FT 1| Alpha(1| Blood group H alpha 2-fucosyltransferase| fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase)| Fucosyltransferase 1| FUT1| FUT1_HUMAN| Galactoside 2 alpha L fucosyltransferase| Galactoside 2-alpha-L-fucosyltransferase 1| GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1| H| HH| HSC| Para Bombay phenotype
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19526
- P19526
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human FUT1
- Synthetic peptide of Human FUT1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together