Anti-PAK2 (phospho-Ser141) Rabbit Polyclonal Antibody
Catalog # 77848-546
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q13177
- Antigen synonyms:p21-activated kinase 2|C-t-EC 2.7.11.1|PAK 2|PAKgamma|Serine threonine protein kinase PAK 2|p34|p58|p21CDKN1A activated kinase 2|PAK-2|p65PAK|Gamma PAK|p21 (CDKN1A)-activated kinase 2a|Serine/threonine protein kinase PAK 2|p21 protein (Cdc42/Rac)-activated kinase 2|p21-activated protein kinase I|S6 H4 kinase|p21-activated kinase|p21 activated kinase 2|p21 protein Cdc42 Rac activated kinase 2|S6/H4 kinase|hPAK65|p21 (CDKN1A) activated kinase 2|PAK-2p34|Gamma-PAK|p27|Kinase|65-KD|PAK2_HUMAN|CB422|PAK65
- Modification:phospho-Ser141
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of serine 141 (Y-L-S(p)-F-T) derived from Human PAK2.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: PAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat