Anti-NENF Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NENF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-999
- 77844-998
- Antigen symbol:
- NENF
- NENF
- Antigen synonyms:
- Cell growth inhibiting protein 47|Cell immortalization-related protein 2|CIR2|NENF_HUMAN|Neudesin|Neuron derived neurotrophic factor|Neuron-derived neurotrophic factor|SCIRP10|SCIRP10 related protein|Secreted protein of unknown function|Spinal cord injury related protein 10|SPUF|SPUF protein
- Cell growth inhibiting protein 47|Cell immortalization-related protein 2|CIR2|NENF_HUMAN|Neudesin|Neuron derived neurotrophic factor|Neuron-derived neurotrophic factor|SCIRP10|SCIRP10 related protein|Secreted protein of unknown function|Spinal cord injury related protein 10|SPUF|SPUF protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UMX5
- Q9UMX5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Neudesin protein (149-167AA)
- Peptide sequence from Human Neudesin protein (149-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together