Anti-RAD54L2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: RAD54L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-891
- 77845-892
- Antigen symbol:
- RAD54L2
- RAD54L2
- Antigen synonyms:
- Androgen receptor interacting protein 4||Helicase ARIP4|ARIP4_HUMAN|SRISNF2L|RAD54-like protein 2|RAD54 like 2 (S. cerevisiae)|HSPC325|Arip4|Androgen receptor-interacting protein 4|HCMV interacting protein|RAD54 like protein 2|Rad54l2|Steroid receptor interacting SNF2 domain protein
- Androgen receptor interacting protein 4||Helicase ARIP4|ARIP4_HUMAN|SRISNF2L|RAD54-like protein 2|RAD54 like 2 (S. cerevisiae)|HSPC325|Arip4|Androgen receptor-interacting protein 4|HCMV interacting protein|RAD54 like protein 2|Rad54l2|Steroid receptor interacting SNF2 domain protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4B4
- Q9Y4B4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Helicase ARIP4 protein (1122-1296AA)
- Recombinant Human Helicase ARIP4 protein (1122-1296AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together