Anti-GSDME Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-127
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GSDME
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60443
- Antigen synonyms:2310037D07Rik|4932441K13Rik|Deafness, autosomal dominant 5|Deafness, autosomal dominant 5 protein|DFNA5|DFNA5 gene|DFNA5_HUMAN|Dfna5h|EG14210|Fin15|ICERE 1|ICERE-1|Inversely correlated with estrogen receptor expression 1|Non-syndromic hearing impairment protein 5|Nonsyndromic hearing impairment protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Gasdermin-E protein (1-496AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: GSDME
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human