Anti-BFAR Rabbit Recombinant Polyclonal Antibody
Catalogue # 77851-863
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:BFAR
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q9NZS9
- Antigen synonyms:BAR|RNF47Bifunctional apoptosis regulator|RING finger protein 47
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Bifunctional apoptosis regulator protein (1-140AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: BFAR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human