Anti-MCM4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-592
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MCM4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P33991
- Antigen synonyms:19G|AI325074|AU045576|CDC 21|CDC 54|CDC21|CDC21 homolog|S. pombe|homolog of|CDC54|Cell division cycle 21|DNA replication licensing factor MCM 4|DNA replication licensing factor hCdc 21|hCdc21|Homolog of S. pombe cell devision cycle 21|KIAA4003|mcdc21|MCM 4|MCM 4 minichromosome maintenance deficient 4|MCM4 minichromosome maintenance deficient 4|mitotin (S. cerevisiae)|MCM4_HUMAN|Mcmd4|MGC33310|Minichromosome maintenance 4|Minichromosome maintenance complex component 4|Minichromosome maintenance deficient (S. cerevisiae) 4|Minichromosome maintenance deficient 4|Minichromosome maintenance deficient 4 homolog (S. cerevisiae)|Minichromosome maintenance|S. cerevisiae|4|mKIAA4003|NKCD|NKGCD|P1 CDC21|P1-CDC21|P1CDC21
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA replication licensing factor MCM4 protein (2-141AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MCM4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat