Anti-BTRC Rabbit Recombinant Polyclonal Antibody
Catalog # 77846-277
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:BTRC
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y297
- Antigen synonyms:bTrCP|Slimb|betaTrCP|BETA-TRCP|beta TrCP1|pIkappaBalpha-E3 receptor subunit|E3RSIkappaB|F box and WD repeats protein beta TrCP|bTrCP1|Beta transducin repeat containing E3 ubiquitin protein ligase|btrC|HOS|F box and WD repeat protein 1B|FBXW1|mKIAA4123|beta-transducin repeat containing|FBW1A|FBXW1A|F-box/WD repeat-containing protein 1A|FBW1A_HUMAN|F-box and WD repeats protein beta-TrCP|F-box/WD repeat protein 1A|FWD1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human F-box/WD repeat-containing protein 1A protein (397-448AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:2000 to 1:10000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: BTRC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human