Anti-EPHA1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-785
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:EPHA1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P21709
- Antigen synonyms:EPH tyrosine kinase|hEpha1|Soluble EPHA1 variant 1|Soluble EPHA1 variant 2|Ephrin receptor Eph A1|EPHT 1|EPH receptor A1|EPH tyrosine kinase 1|Erythropoietin-producing hepatoma receptor|MGC163163|Oncogene EPH|Esk|EPHT|Tyrosine-protein kinase receptor EPH|Epha1|EPHT1|Ephrin type A receptor 1|Erythropoietin producing hepatoma amplified sequence|EPHA1_HUMAN|Tyrosine protein kinase receptor EPH|Ephrin type-A receptor 1|EPH
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ephrin type-A receptor 1 protein (296-414AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: EPHA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse