Anti-IFNG Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-901
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:IFNG
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P01579
- Antigen synonyms:IF 1|IFG|IFI|IFN gamma|IFN immune|IFN, immune|IFN-gamma|IFNG_HUMAN|Immune interferon|Interferon gamma|Type II Interferon
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Interferon gamma protein (24-162AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: IFNG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human