Anti-APPL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: APPL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-047
- 77844-046
- Antigen symbol:
- APPL1
- APPL1
- Antigen synonyms:
- Adapter protein containing PH domain|Adaptor Protein Containing pH Domain|Adaptor protein containing pH domain PTB domain and leucine zipper motif 1|Adaptor protein containing pH domain PTB domain and leucine zipper motif|Adaptor protein phosphotyrosine interacting with PH domain and leucine zipper 1|AKT2 INTERACTOR|APPL 1|Appl1|DCC interacting protein 13 alpha|DCC-interacting protein 13-alpha|DIP 13 alpha|DIP13 alpha|Dip13-alpha|DIP13alpha|DP13A_HUMAN|KIAA1428|PTB domain and leucine zipper motif 1|PTB Domain and Leucine Zipper Motif|Signaling adaptor protein DIP13alpha
- Adapter protein containing PH domain|Adaptor Protein Containing pH Domain|Adaptor protein containing pH domain PTB domain and leucine zipper motif 1|Adaptor protein containing pH domain PTB domain and leucine zipper motif|Adaptor protein phosphotyrosine interacting with PH domain and leucine zipper 1|AKT2 INTERACTOR|APPL 1|Appl1|DCC interacting protein 13 alpha|DCC-interacting protein 13-alpha|DIP 13 alpha|DIP13 alpha|Dip13-alpha|DIP13alpha|DP13A_HUMAN|KIAA1428|PTB domain and leucine zipper motif 1|PTB Domain and Leucine Zipper Motif|Signaling adaptor protein DIP13alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKG1
- Q9UKG1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DCC-interacting protein 13-alpha protein (214-495AA)
- Recombinant Human DCC-interacting protein 13-alpha protein (214-495AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together