Anti-ASAH2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77851-455
New Product
- Antigen symbol:ASAH2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NR71
- Antigen synonyms:Acylsphingosine deacylase 2| AI585898| ASAH2| ASAH2_HUMAN| BCDase| hCD| HNAC1| LCDase| MGC129777| mitochondrial ceramidase| N acylsphingosine amidohydrolase (non lysosomal ceramidase) 2| N acylsphingosine amidohydrolase 2| N CDase| N-acylsphingosine amidohydrolase 2| N-CDase| NCDase| Neutral ceramidase soluble form| Non lysosomal ceramidase| Non-lysosomal ceramidase
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Neutral ceramidase protein (255-506AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ASAH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human