Anti-TRAF3IP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: TRAF3IP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77849-137
- 77849-136
- Antigen symbol:
- TRAF3IP1
- TRAF3IP1
- Antigen synonyms:
- Interleukin 13 receptor alpha 1 binding protein 1|Interleukin-13 receptor alpha 1-binding protein 1|Microtubule interacting protein that associates with TRAF3|Microtubule-interacting protein associated with TRAF3|MIP T3|MIP-T3|MIPT3|MIPT3_HUMAN|TNF receptor associated factor 3 interacting protein 1|TRAF3-interacting protein 1|Traf3ip1
- Interleukin 13 receptor alpha 1 binding protein 1|Interleukin-13 receptor alpha 1-binding protein 1|Microtubule interacting protein that associates with TRAF3|Microtubule-interacting protein associated with TRAF3|MIP T3|MIP-T3|MIPT3|MIPT3_HUMAN|TNF receptor associated factor 3 interacting protein 1|TRAF3-interacting protein 1|Traf3ip1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDR0
- Q8TDR0
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TRAF3IP1
- Fusion protein of Human TRAF3IP1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together