Anti-AHCY Rabbit Recombinant Polyclonal Antibody (Biotin)
77850-912
New Product
- Antigen symbol:AHCY
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P23526
- Antigen synonyms:Adenosylhomocysteinase|AdoHcyase|ahcY|S adenosyl L homocysteine hydrolase|S adenosylhomocysteine hydrolase|S-adenosyl-L-homocysteine hydrolase|SAHH|SAHH_HUMAN
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenosylhomocysteinase protein (2-432AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: AHCY
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human