Anti-CNOT1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-335
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CNOT1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:A5YKK6
- Antigen synonyms:CDC39|KIAA1007|NOT1|AD-005CCR4-NOT transcription complex subunit 1|CCR4-associated factor 1|Negative regulator of transcription subunit 1 homolog|NOT1H|hNOT1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human CCR4-NOT transcription complex subunit 1 protein (604-867AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CNOT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human