Order Entry
Canada
ContactUsLinkComponent
Anti-KLF12 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-KLF12 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-KLF12 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77846-396
  • 77846-397
  • Antigen symbol:
  • KLF12
  • KLF12
  • Antigen synonyms:
  • 2700063E05Rik|Transcriptional repressor AP 2rep|Krueppel-like factor 12|Transcriptional repressor AP-2rep|AP2REP|D530033K05Rik|KLF12_HUMAN|Krueppel like factor 12|AP-2rep transcription factor|KLF12 zinc finger transcriptional repressor|AI315654|OTTHUMP00000018503|AI225908|AP-2rep|KLF12|HSPC122|AP-2 repressor|B130052C06Rik
  • 2700063E05Rik|Transcriptional repressor AP 2rep|Krueppel-like factor 12|Transcriptional repressor AP-2rep|AP2REP|D530033K05Rik|KLF12_HUMAN|Krueppel like factor 12|AP-2rep transcription factor|KLF12 zinc finger transcriptional repressor|AI315654|OTTHUMP00000018503|AI225908|AP-2rep|KLF12|HSPC122|AP-2 repressor|B130052C06Rik
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y4X4
  • Q9Y4X4
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Krueppel-like factor 12 protein (36-313AA)
  • Recombinant Human Krueppel-like factor 12 protein (36-313AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together