Anti-ARID1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-375
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ARID1A
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O14497
- Antigen synonyms:actin-dependent regulator of chromatin subfamily F member 1|ARI1A_HUMAN|ARID domain containing protein 1A|ARID domain-containing protein 1A|ARID1A|AT rich interactive domain 1A (SWI like)|AT rich interactive domain 1A|AT rich interactive domain containing protein 1A|AT-rich interactive domain-containing protein 1A|B120|BAF250|BAF250A|BM029|brain protein 120|BRG1 associated factor 250|BRG1 associated factor 250a|BRG1-associated factor 250|BRG1-associated factor 250a|C1ORF4|chromatin remodeling factor p250|chromosome 1 open reading frame 4|ELD|hELD|hOSA1|matrix-associated|MRD14|Osa homolog 1|OSA1|OSA1 nuclear protein|P270|SMARCF1|SWI like protein|SWI SNF complex protein p270|SWI-like protein|SWI/SNF complex protein p270|SWI/SNF related|matrix associated|actin dependent regulator of chromatin|subfamily f|member 1|SWI/SNF-related
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human AT-rich interactive domain-containing protein 1A protein (655-816AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ARID1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human