Anti-CLN5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-262
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CLN5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75503
- Antigen synonyms:Ceroid lipofuscinosis neuronal 5|Ceroid-lipofuscinosis neuronal protein 5|CLN5_HUMAN|NCL|Protein CLN5
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ceroid-lipofuscinosis neuronal protein 5 protein (42-181AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: CLN5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human