Anti-CD46 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: CD46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-837
- 77847-836
- Antigen symbol:
- CD46
- CD46
- Antigen synonyms:
- AHUS2|Antigen defined by monoclonal TRA 2 10|Antigen identified by monoclonal TRA 2 10|CD46|CD46 antigen|CD46 antigen complement regulatory protein|CD46 molecule|CD46 molecule complement regulatory protein|Complement membrane cofactor protein|MCP|MCP_HUMAN|Measles virus receptor|membrane cofactor protein (CD46|trophoblast-lymphocyte cross-reactive antigen)|Membrane cofactor protein|MGC26544|MIC10|TLX|TRA2.10|Trophoblast leucocyte common antigen|Trophoblast leukocyte common antigen|Trophoblast lymphocyte cross reactive antigen
- AHUS2|Antigen defined by monoclonal TRA 2 10|Antigen identified by monoclonal TRA 2 10|CD46|CD46 antigen|CD46 antigen complement regulatory protein|CD46 molecule|CD46 molecule complement regulatory protein|Complement membrane cofactor protein|MCP|MCP_HUMAN|Measles virus receptor|membrane cofactor protein (CD46|trophoblast-lymphocyte cross-reactive antigen)|Membrane cofactor protein|MGC26544|MIC10|TLX|TRA2.10|Trophoblast leucocyte common antigen|Trophoblast leukocyte common antigen|Trophoblast lymphocyte cross reactive antigen
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15529
- P15529
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CD46
- Fusion protein of Human CD46
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together