Recombinant Human PR3 (from HEK293 cells)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Frequently Bought Together
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Water, deionized VWR®
Supplier: VWR
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Single-Use Needles, BD Medical
Supplier: BD
Advanced® Bilirubin STAT-Analyzer™, Model BR2, Advanced Instruments
Supplier: Advanced Instruments
AGAR BRAIN HEART INFUSION 450 G
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex